Learn More
Biolegend PE/DazzleTM 594 anti-mouse CD6
Supplier:  Biolegend 161216
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD62L [W18021D]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Catalog No. 50-272-1538