Learn More
Biolegend PE/Dazzle™ 594 anti-human IgD
Supplier:  Biolegend 348239
Encompass_Preferred
PE/Dazzle™ 594 anti-human IgD [IA6-2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-6868