Learn More
Medchemexpress LLC Phycoerythrin-conjugated recombinant human glypican-3 (His-tag) | 50UG
SDP
Supplier:  Medchemexpress LLC HYP70129350UG
Encompass_Preferred
PE-labeled Glypican-3 (GPC3) is a recombinant human extracellular proteoglycan produced in HEK293 cells and conjugated to phycoerythrin for use as a fluorescent detection reagent. The protein includes a His tag and is supplied as a purified, labeled protein suitable for immunoassays, flow cytometry, and antibody validation workflows.
  • Phycoerythrin-conjugated recombinant human GPC3 for fluorescent detection.
  • Expressed in HEK293 cells to preserve native post-translational modifications.
  • C-terminal His tag for reliable purification and detection.
  • Ready-to-use reagent for flow cytometry and binding assays.
  • Supplied as a purified, labeled protein for antibody validation and assay development.
  • Store and handle per recommended conditions to maintain activity.
Catalog No. 50-001-76331