Learn More
Medchemexpress LLC PE-labeled HLA-G&B2M&peptide (RIIPRHLQL) tetramer protein, human (HEK293, His-Avi) | 100UG
SDP
Supplier:  Medchemexpress LLC HYP701002100UG
Encompass_Preferred
PE-labeled HLA-G&B2M&Peptide (RIIPRHLQL) tetramer protein supplied as a lyophilized reagent for immunological assays. The reagent combines human HLA-G heavy chain, beta-2 microglobulin, and the RIIPRHLQL peptide, expressed in HEK293 cells and conjugated to phycoerythrin to enable direct fluorescent detection in binding and cell-staining applications.
Catalog No. 50-001-76206