Learn More
ACROBiosystems ACROBiosystems PE-Labeled Human CD4 Protein, His Tag (Site-specific conjugation)
Supplier:  ACROBiosystems CD4HP2E325TESTS
Encompass_Preferred
Recombinant Protein;25TESTS;PE-Labeled Human CD4, His Tag (CD4-HP2E3) is produced via site-specific conjugation of PE to Human CD4, His Tag under optimal conditions with a proprietary technology. Human CD4, His Tag is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Pro 396 (Accession # P01730-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45.0 kDa. ;5e5 of anti-CD4 CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) of PE-Labeled Human CD4, His Tag (Cat. No. CD4-HP2E3) and negative control protein respectively. PE signal was used to evaluate the binding activity (QC tested).
Catalog No. 50-210-6559