Learn More
ACROBiosystems ACROBiosystems PE-Labeled Human CLEC12A / MICL / CLL-1 Protein, His Tag (Site-specific conjugation)
Supplier:  ACROBiosystems CLAHP2Q325TESTS
Encompass_Preferred
Recombinant Protein;25TESTS;PE-Labeled Human CLEC12A, His Tag (CLA-HP2Q3) is produced via site-specific conjugation of PE to Human CLEC12A, His Tag under optimal conditions with a proprietary technology. Human CLEC12A, His Tag is expressed from human 293 cells (HEK293). It contains AA His 65 - Ala 265 (Accession # Q5QGZ9-2).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 27.3 kDa. ;5e5 of anti-CLEC12A/CLL-1 CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) of PE-Labeled Human CLEC12A, His Tag (Cat. No. CLA-HP2Q3) and negative control protein respectively. PE signal was used to evaluate the binding activity (QC tested).
Catalog No. 50-210-6593