Learn More
ABclonal Technology PE Mouse anti-Human CD56 mAb
SDP
Supplier:  ABclonal Technology A26868500T
Encompass_Preferred
This gene encodes a cell adhesion protein in the immunoglobulin superfamily involved in cell-to-cell and cell-matrix interactions during development It regulates neurogenesis neurite outgrowth and cell migration in the nervous system The protein also aids in the expansion of T lymphocytes B lymphocytes and natural killer (NK) cells essential for immune surveillance It interacts with fibroblast growth factor receptors and other extracellular matrix components triggering signaling pathways such as FYN-FAK MAPK and PI3K The CD56 isoform plays a role in myeloproliferative disorders like acute myeloid leukemia with its expression linked to disease progression and lower survival rates Increased CD56 expression is also associated with severe COVID-19 Alternative splicing results in multiple isoforms
Catalog No. 50-000-08476