Learn More
ABclonal Technology PE Rabbit anti-Human CD94 mAb
SDP
Supplier:  ABclonal Technology A26686100T
Encompass_Preferred
Natural killer (NK) cells are a distinct lineage of lymphocytes that mediate cytotoxic activity and secrete cytokines upon immune stimulation Several genes of the C-type lectin superfamily including members of the NKG2 family are expressed by NK cells and may be involved in the regulation of NK cell function KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus Three transcript variants encoding two different isoforms have been found for this gene
Catalog No. 50-000-12001