Learn More
ABclonal Technology PE Rabbit anti-Mouse CD200/OX2
SDP
Supplier:  ABclonal Technology A26859100T
Encompass_Preferred
Predicted to enable protein binding activity involved in heterotypic cell-cell adhesion Involved in negative regulation of cell population proliferation negative regulation of macrophage activation and negative regulation of neuroinflammatory response Located in cell surface Is expressed in several structures including anterior naris aorta bone nervous system and retina Used to study autoimmune disease Orthologous to human CD200 (CD200 molecule)
Catalog No. 50-000-08524