Learn More
ABclonal Technology PE Rabbit anti-Mouse CD22 mAb
SDP
Supplier:  ABclonal Technology A25960100T
Encompass_Preferred
Enables CD4 receptor binding activity protein phosphatase binding activity and sialic acid binding activity Involved in several processes including negative regulation of calcium-mediated signaling negative regulation of immunoglobulin production and regulation of B cell proliferation Located in external side of plasma membrane and neuronal cell body membrane Is integral component of plasma membrane Is expressed in several structures including alimentary system brain hemolymphoid system gland liver and biliary system and reproductive system Orthologous to human CD22 (CD22 molecule)
Catalog No. 50-000-13163