Learn More
ABclonal Technology PE Rabbit anti-Mouse CD23 mAb
SDP
Supplier:  ABclonal Technology A27347100UG
Encompass_Preferred
Predicted to enable carbohydrate binding activity Acts upstream of or within positive regulation of humoral immune response mediated by circulating immunoglobulin Located in external side of plasma membrane Orthologous to human FCER2 (Fc fragment of IgE receptor II)
Catalog No. 50-000-10409