Learn More
ABclonal Technology PE Rabbit anti-Mouse CD25 mAb
SDP
Supplier:  ABclonal Technology A26508200T
Encompass_Preferred
Enables interleukin-2 binding activity and interleukin-2 receptor activity Acts upstream of or within several processes including Notch signaling pathway activation-induced cell death of T cells and regulation of T cell proliferation Located in external side of plasma membrane Is expressed in bone marrow Used to study Sjogren s syndrome and inflammatory bowel disease Human ortholog(s) of this gene implicated in immunodeficiency 41 lymphopenia multiple sclerosis type 1 diabetes mellitus and type 1 diabetes mellitus 10 Orthologous to human IL2RA (interleukin 2 receptor subunit alpha)
Catalog No. 50-000-12800