Learn More
ABclonal Technology PE Rabbit anti-Mouse CD32 mAb
SDP
Supplier:  ABclonal Technology A26142100T
Encompass_Preferred
CD32 (Fcgr2) is a 40 kD transmembrane glycoprotein member of the immunoglobulin superfamily The extracellular region of CD32 consists of two Ig C-type domains that binds the Fc region from monomeric IgG with low affinity but binds immune complexes efficiently CD32 can mediate phagocytosis of immune complexes and modulate cell activation CD32 is expressed by Macrophages neutrophils mast cells and B cells
Catalog No. 50-000-09215