Learn More
ABclonal Technology PE Rabbit anti-Mouse CD69 mAb
SDP
Supplier:  ABclonal Technology A26445500T
Encompass_Preferred
Predicted to enable calcium ion binding activity and identical protein binding activity Located in external side of plasma membrane Orthologous to human CD69 (CD69 molecule)
Catalog No. 50-000-11613