Learn More
ABclonal Technology PE Rabbit anti-Mouse CD88/C5aR
SDP
Supplier:  ABclonal Technology A26709100T
Encompass_Preferred
Enables complement component C5a receptor activity Involved in several processes including astrocyte activation positive regulation of vascular endothelial growth factor production and regulation of granulocyte chemotaxis Acts upstream of or within defense response to Gram-positive bacterium neutrophil chemotaxis and response to peptidoglycan Predicted to be located in several cellular components including apical part of cell basolateral plasma membrane and cell surface Predicted to be active in plasma membrane Is expressed in several structures including brain liver neural ectoderm and sensory organ Orthologous to several human genes including OPRPN (opiorphin prepropeptide)
Catalog No. 50-000-12901