Learn More
ABclonal Technology PE Rabbit anti-Mouse FOXP3 mAb
SDP
Supplier:  ABclonal Technology A26227500T
Encompass_Preferred
The protein encoded by this gene is a member of the forkhead winged-helix family of transcriptional regulators Defects in this gene result in the scurfy phenotype (sf) Alternative splicing results in multiple transcript variants
Catalog No. 50-000-08810