Learn More
ABclonal Technology PE Rabbit anti-Mouse IL-13 mAb
SDP
Supplier:  ABclonal Technology A26687200T
Encompass_Preferred
Predicted to enable interleukin-13 receptor binding activity Involved in positive regulation of cold-induced thermogenesis and positive regulation of macrophage activation Acts upstream of or within cellular response to cytokine stimulus inflammatory response and positive regulation of immune effector process Located in cytoplasm external side of plasma membrane and extracellular space Is expressed in gut liver male reproductive gland or organ and thymus Used to study atopic dermatitis Human ortholog(s) of this gene implicated in several diseases including allergic disease (multiple) autoimmune disease (multiple) diffuse scleroderma lung disease (multiple) and nose disease (multiple) Orthologous to human IL13 (interleukin 13)
Catalog No. 50-000-12008