Learn More
Cytek Biosciences Inc PE Rt IgG2a Iso Con Anti Isoty
Supplier: Cytek Biosciences Inc 504321U100
Product Name: PE Rat IgG2a Isotype Control (2A3)
Product Class: Antibody
Type: Isotype Control
Product Name: PE Rat IgG2a Isotype Control (2A3)
Cytek Catalog Number: 50-4321-U100
Marker: Isotype Control
Clonality: Monoclonal
Clone: 2A3
Host Species: Rat
Isotype: IgG2a
Conjugation: PE
Size: 100 μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.