Learn More
MSC PIP CoreShield Cut Resistant Gloves, 18 Gauge, Size 10, 1/Pr.
Supplier: MSC 37094802
These 18-gauge cut-resistant gloves provide advanced protection and dexterity for precision tasks in demanding industrial environments. Designed for comfort and high performance, they feature a specialized liner and a dual-layer nitrile coating to ensure grip and durability.
- Seamlessly knitted liner made of recycled polyester, steel, and PE fiber
- Double coating with 3/4 smooth nitrile and nitrile micro-foam palm
- Cut protection level A/A2 and B
- Reinforced thumb area for added longevity
- Fiberglass-free and silicone-free construction
- Antistatic properties certified according to EN 16350
- Touchscreen-capable for ease of use with electronic devices
- Food certified for safe handling in relevant applications
- Color-coded for efficient size identification