Learn More
MSC PIP Cut Resistant Gloves, HPPE, 18 Gauge, Size M, 1/Pr.
Supplier:  MSC 37110384
Encompass
These size M gloves are engineered for high-performance protection and versatility in demanding industrial settings. Constructed with an 18-gauge, fiberglass-free liner, the gloves provide excellent dexterity and tactile sensitivity while maintaining A5 cut resistance. The palm and fingers feature a durable, breathable foam nitrile coating designed to disperse fluids for a superior grip in wet or dry conditions.
  • Seamless knit construction enhances wearer comfort and breathability.
  • Reinforced thumb crotch improves cut resistance and extends the service life in high-wear areas.
  • Touchscreen compatible design allows for device operation without removing gloves.
  • ESD and silicone-free construction for specialized handling tasks.
  • Compliant with 21 CFR standards for food contact.
  • Machine washable and reusable for cost-effective maintenance.
Catalog No. 50-005-08603