Learn More
MSC PIP CoreShield Cut Resistant Gloves, A4, 18 Gauge, White, 10 Pair
Supplier:  MSC 37107299
Encompass
These CoreShield gloves are designed for high-level cut protection and enhanced durability. Featuring an 18-gauge construction, they offer a lightweight and breathable design suitable for handling small parts while maintaining superior grip and touch screen compatibility. The construction utilizes an engineered yarn that is free from glass fiber, ensuring a comfortable fit for extended use.
  • Provides ANSI/ISEA 105 cut level A4 protection
  • Features a durable nitrile foam coating for high abrasion resistance
  • Includes reinforced thumb design for increased longevity
  • Certified for food handling applications
  • Silicone-free and antistatic properties
  • Equipped with a color-coded sizing system for easy identification
  • Low content level of DMF (<100 ppm)
  • Fully washable for repeated use
Catalog No. 50-005-08180