Learn More
MSC PIP CoreShield™ 3.0 Cut-Resistant Gloves, Nitrile Palm, A8, 18 ga, 2X-Large, 10/Pk.
Supplier: MSC 37113198
These CoreShield™ 3.0 cut-resistant gloves provide superior hand protection in demanding industrial environments. Engineered with a lightweight, seamless 18-gauge liner and nitrile foam palm coating, they offer high dexterity, tactile sensitivity, and excellent grip in dry, wet, and oily conditions. The design is reinforced between the thumb and forefinger for added durability and includes ESD-safe construction for specialized applications.
- ANSI/ISEA cut level A8 and EN 388:2016 4X42F protection
- Reinforced thumb crotch for extended wear
- Touchscreen compatible and silicone-free design
- Certified for food handling per 21 CFR 177.2600
- Electrostatic properties compliant with EN 16350
- Reusable and machine washable at 40°C
- Oeko-Tex® standard 100 certified
- Knit wrist cuff keeps out debris