Learn More
MSC PIP CoreShield 3.0 Cut-Resistant Gloves, A8, 18 ga, Size Large, 100/Case
Supplier:  MSC 37113685
Encompass
These seamless knit gloves provide advanced cut protection with an ANSI cut level A8. Constructed from a lightweight 18-gauge fiberglass-free yarn, they offer superior tactile sensitivity, dexterity, and comfort for extended wear. The foam nitrile coating on the palm and fingers features a cell structure designed to disperse fluids on contact, ensuring a reliable grip in dry, wet, and oily conditions.
  • Provides ANSI cut level A8 protection
  • Features a reinforced thumb crotch to extend glove life
  • Touchscreen compatible for device operation
  • ESD safe construction for use with static-sensitive materials
  • Food certified
  • Machine washable for reusability
  • Silicone free
  • Length: 9.83 in. with a 3 in. cuff
Catalog No. 50-005-08988