Learn More
MSC PIP Glove, HPPE, Nitrile Coated, 18 Gauge, Size 9, 1/Pr.
Supplier:  MSC 37111408
Encompass
These G-Tek® CoreShield® gloves provide high-level cut resistance and dexterity for demanding environments. Constructed with an 18-gauge, fiberglass-free HPPE liner, they offer a comfortable, lightweight fit with enhanced tactile sensitivity. The black foam nitrile coating on the palm and fingers features a unique cell structure that disperses fluids to maintain a reliable grip in various conditions.
  • ANSI cut level A6 and puncture level 3 for superior hand protection.
  • Touchscreen compatible design allows for device operation without glove removal.
  • Reinforced thumb crotch improves durability and cut resistance in high-wear areas.
  • ESD safe construction suitable for sensitive electronic applications.
  • Certified for food contact and launderable for extended service life.
Catalog No. 50-005-08984