Learn More
MSC G-Tek CoreShield Engineered Yarn Glove, Nitrile Foam Coated, Cut Level A9, Size 10, 100/Case
Supplier: MSC 37114212
These vend-ready gloves feature a seamless knit construction with a high-performance engineered yarn liner, designed to provide superior cut resistance and comfort in industrial settings. The 18-gauge, fiberglass-free liner ensures tactile sensitivity, while the durable foam nitrile coating on the palm and fingers offers an excellent grip in wet or oily conditions.
- Provides A9 cut level protection and ANSI level 3 abrasion and puncture resistance
- Features a reinforced thumb crotch to extend service life in high-stress areas
- Constructed with a breathable, touchscreen-compatible material for ease of use
- Certified for food contact and silicone-free for versatile application
- Designed for compatibility with personal protective equipment vending machines
- Reusable and launderable for extended service life