Learn More
Medchemexpress LLC Peanut agglutinin | ≥85% | 1 MG
Supplier: Medchemexpress LLC HYP34431MG
Peanut agglutinin (PNA) is a plant lectin. It functions as a probe to specifically bind biomolecules like polysaccharides and peptides. This biological material or organic compound is suitable for use in life science research.
- Solid appearance
- White to off-white color
- Initial source: plants Leguminosae Arachis hypogaea L.
- Functions as a probe to specifically bind biomolecules (such as polysaccharides, peptides, etc.)
- Suitable for life science research