Learn More
Medchemexpress LLC Peanut Agglutinin (Fluorescein) | 1 MG
Supplier: Medchemexpress LLC HYNP01381MG
Peanut Agglutinin (PNA) Fluorescein is a plant lectin used as a probe to specifically bind biomolecules such as polysaccharides and peptides. It is a biological material or organic compound suitable for life science research.
- Used as a probe to specifically bind biomolecules
- Biological material for life science research
- Organic compound for life science research