Learn More
Medchemexpress LLC Peanut Agglutinin (Rhodamine) | 1 MG
SDP
Supplier:  Medchemexpress LLC HYNP01561MG
Encompass_Preferred
Peanut Agglutinin (PNA) Rhodamine is a plant lectin serving as a probe for specific binding to biomolecules such as polysaccharides and peptides. It is a biological material or organic compound well-suited for various life science research applications.
  • Plant lectin
  • Functions as a probe for biomolecule binding
  • Binds to polysaccharides, peptides, and other biomolecules
  • Biological material or organic compound
  • Suitable for life science research
Catalog No. 50-004-25340