Learn More
ABclonal Technology PEBP4 Rabbit pAb
SDP
Supplier:  ABclonal Technology A144851000UL
Encompass_Preferred
The phosphatidylethanolamine (PE)-binding proteins, including PEBP4, are an evolutionarily conserved family of proteins with pivotal biologic functions, such as lipid binding and inhibition of serine proteases (Wang et al., 2004 [PubMed 15302887]).
Catalog No. 50-001-06759