Learn More
Medchemexpress LLC Pebrellin | 13509-93-8 | 374.34 | 5 MG

Supplier: Medchemexpress LLC HYN127345MG
Pebrellin is a flavonoid that can be isolated from *Mentha x piperita* leaves. It is for research use only and not sold to patients.
- Flavonoid isolated from Mentha x piperita leaves
- For research use only
- Purity: 98.51%
- Appearance: Solid
- Color: Light yellow to yellow
- Structure classification: Flavonoids, Flavones
- Initial source: Plants (Leguminosae, *Acacia farnesiana (Linn.) Willd.*), Plants (Labiatae)
- Related to inflammation or immune system disease
- Natural product standard
- Inhibitor
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.