Learn More
Medchemexpress LLC HY-N0493 5mg Medchemexpress, Pectolinarigenin CAS:520-12-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN04935MG
Encompass_Preferred
Medchemexpress, HY-N0493 5mg Pectolinarigenin CAS:520-12-7 Pectolinarigenin, isolated from Cirsium chanroenicum, is a dual inhibitor of COX-2/5-LOX. Anti-inflammatory activity. Pectolinarigenin has potent inhibitory activities on melanogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2794