Learn More
Abcam PE/Cy5.5 Conjugation Kit - Lightning-Link, 3 x 10UG
Supplier:  Abcam AB1028993X10UG
Encompass_Preferred
Labeling in < 4 hrs with 30 secs hands-on time using PE/Cy5.5 Conjugation Kit - Lightning-Link ab102899. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5594