Learn More
Perkin Elmer US LLC PEEK Finger-tight Tubing Nut for GemClean Nebulizer
Supplier:  Perkin Elmer US LLC 09920516
Encompass_Preferred
This PEEK finger-tight tubing nut is used on the sample tube connection to GemClean Cross-Flow II nebulizers of ICP-MS instruments.
Catalog No. 50-211-3092