Learn More
Trajan Scientific PEEK TUBE NATURAL 1/16” OD X 0.03” ID, LENGTH 50 FEET
SDP
Supplier:  Trajan Scientific 130203000050F
Encompass_Preferred
PEEK TUBE NATURAL 1/16” OD X 0.03” ID, LENGTH 50 FEET
Catalog No. 50-272-4811