Learn More
Medchemexpress LLC Peficitinib | 944118-01-8 | 99.5% | 326.39 g/mol | C18H22N4O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY19568R10MG
Encompass_Preferred
Peficitinib (Standard) is an analytical reference standard of peficitinib, an orally active Janus kinase (JAK) inhibitor supplied as a high-purity solid for research use. It is intended for method development, calibration, and qualitative or quantitative analysis by HPLC, GC, and MS.
  • High purity: reported 99.5%.
  • Suitable for HPLC, GC, and MS analytical methods.
  • Molecular formula C18H22N4O2; molecular weight 326.39 g/mol.
  • Supplied as a solid ideal for method development and calibration.
  • Available in small analytical pack sizes such as 10 MG for standards work.
  • Identified by CAS number 944118-01-8 for unambiguous reference.
Catalog No. 50-002-98313