Learn More
Medchemexpress LLC Pefloxacin | 70458-92-3 | 99.4% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB01475MG
Encompass_Preferred
Pefloxacin is a broad-spectrum antibiotic that functions by inhibiting DNA gyrase, thereby blocking DNA replication. It also inhibits DNA relaxation catalyzed by topoisomerase I with an IC₅₀ of 45 μg/mL. This antibiotic exhibits activity against various bacteria including *Escherichia coli*, *Pseudomonas aeruginosa*, and *Bacteroides fragilis*, and has anti-*Plasmodium yoelii* infection activity.
  • Antibacterial activity against various bacteria.
  • Anti-malarial activity, reducing parasitemia and increasing survival rates.
  • Useful for infection studies.
  • Can increase UVA-induced edema and immunosuppression.
Catalog No. 50-004-50905