Learn More
Medchemexpress LLC Pefloxacin-d5 (pefloxacinium-d5) | 1228182-51-1 | MFCD16652561 | 98.3% | 338.39 | C17H15D5FN3O3 | 10 MG

Supplier: Medchemexpress LLC HYB0147S10MG
Pefloxacin-d5 is the deuterium-labeled analogue of the fluoroquinolone antibiotic pefloxacin, provided as a high-purity stable isotope standard for use as an internal standard in quantitative mass spectrometry, pharmacokinetic, and infection studies.
- High purity (98.3%).
- Chemical formula: C17H15D5FN3O3.
- Molecular weight: 338.39.
- CAS number: 1228182-51-1.
- Suitable for quantitative LC-MS/MS and pharmacokinetic studies.
- 10 mg packaging available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.