Learn More
Apexbio Technology LLC Pefloxacin Mesylate Dihydrate, 10g. Cas: 149676-40-4 MFCD: MFCD01685696. third generation of fluoroquinolone class of antibacterials
SDP
Supplier:  Apexbio Technology LLC B229110000
Encompass_Preferred
Pefloxacin mesylate dehydrate is a an antibacterial agent and prevents bacterial DNA replication by inhibiting DNA gyrase (topoisomerse) Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5339