Learn More
Cayman Chemical PefloxacInmesylate hydrate 25g
SDP
Supplier:  Cayman Chemical 2910325 G
Encompass_Preferred
A broad-spectrum and synthetic fluoroquinolone antibiotic; active against clinical isolates of E. coli, K. pneumoniae, S. aureus, and G. vaginalis (MIC90s = 0.12, 0.5, 0.5, and 32 mg/L, respectively) selectively inhibits E. coli topoisomerase I and DNA gyrase over calf thymus topoisomerase I (IC50s = 45, 4.5, and >140 μg/ml, respectively) in cell-free assays; active against systemic E. coli, K. pneumoniae, and P. aeruginosa infections in mice (P50s = 6.5, 29.2, and 219 mg/kg, respectively)
Catalog No. 50-293-8249