Learn More
Medchemexpress LLC Peg(2000)-C-DMG | 1443687-74-8 | 98.0% | 2600 (Approximately) | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY168399
Encompass_Preferred
PEG(2000)-C-DMG is a PEGylated derivative of 1,2-Dimyristoyl-sn-glycerol. It can be utilized in conjunction with other lipids for the creation of lipid nanoparticles (LNPs) that are used in the delivery of siRNA.
  • Can be used in combination with other lipids for the formation of lipid nanoparticles (LNPs).
  • Useful for the delivery of siRNA.
  • For research use only, not intended for sale to patients.
Catalog No. 50-003-94062