Learn More
Medchemexpress LLC PEG 4 lauryl ether 1g
SDP
Supplier:  Medchemexpress LLC HYW0995361G
Encompass_Preferred
PEG 4 lauryl ether can be used as an excipient Pharmaceutical excipients or pharmaceutical auxiliaries refer to other chemical substances used in the pharmaceutical process other than pharmaceutical ingredients Pharmaceutical excipients generally refer to inactive ingredients in pharmaceutical preparations which can improve the stability solubility and processability of pharmaceutical preparations Pharmaceutical excipients also affect the absorption distribution metabolism and elimination (ADME) processes of co-administered drugs[1]
Catalog No. 50-002-05825