Learn More
Cayman Chemical PEG damne 1g
SDP
Supplier:  Cayman Chemical 21174 1G
Encompass_Preferred
ERRY Reporter Cells are prepared using INDIGOs proprietary CryoMite process This cryo-preservation method yields high cell viability post-thaw and provides the convenience of immediately dispensing healthy division-competent reporter cells into assay plates There is no need for intermediate spin-and-wash steps viability determinations or cell titer adjustments [INDIGO Catalog Nos IB08021-32 IB08021 IB08022]
Catalog No. 50-298-1244