Learn More
Cayman Chemical PEG damne 5g
SDP
Supplier:  Cayman Chemical 21174 5G
Encompass_Preferred
A DYPD inhibitor (Ki 0 36 UM) inhibits the degradation of 5-FU in rat liver extracts (IC50 0 1 UM) inhibits homologous recombination and enhances radiation-induced cell death in DLD-1 human colorectal cancer cells
Catalog No. 50-298-1423