Learn More
Hampton Research PEG/Ion 2 Screen, 10ml
SDP
Supplier:  Hampton Research HR2098
Encompass
PEG/Ion 2 is an extension to the fundamental crystallization strategy in PEG/Ion. PEG/Ion 2 reagents cover the monodisperse, high purity Polyethylene glycol 3,350 and an array of neutralized and pH adjusted organic acids, multivalent ions, a novel Citrate BIS-TRIS propane buffer system and pH (4 - 8.8). Each of the 48 reagents in PEG/Ion 2 contains PEG 3,350 as the polymer (precipitant). The concentration of PEG is varied from 12% w/v to 20% w/v depending upon the type and concentration of buffer/salt paired with the polymer. 10ml Please note items from this supplier are non-returnable.
Catalog No. 50-093-4