Learn More
Hampton Research HR2-998-16/PEG/Ion 2 Screen #16 (F04), 185 ml - 8% v/v Tacsimate pH 7.0, 20% w/v Polyethylene glycol 3,350
SDP
Supplier:  Hampton Research HR299816
Encompass
HR2-998-16/PEG/Ion 2 Screen #16 (F04), 185 ml - 8% v/v Tacsimate pH 7.0, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-246-8