Learn More
Hampton Research HR2-998-17/PEG/Ion 2 Screen #17 (F05), 185 ml - 4% v/v Tacsimate pH 8.0, 12% w/v Polyethylene glycol 3,350
SDP
Supplier:  Hampton Research HR299817
Encompass
HR2-998-17/PEG/Ion 2 Screen #17 (F05), 185 ml - 4% v/v Tacsimate pH 8.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-246-9