Learn More
Hampton Research HR2-998-33/PEG/Ion 2 Screen #33 (G09), 185 ml - 2% v/v Tacsimate pH 6.0, 0.1 M BIS-TRIS pH 6.5, 20% w/v Polyethylene glycol 3,350
SDP
Supplier:  Hampton Research HR299833
Encompass
HR2-998-33/PEG/Ion 2 Screen #33 (G09), 185 ml - 2% v/v Tacsimate pH 6.0, 0.1 M BIS-TRIS pH 6.5, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-248-5