Learn More
Hampton Research HR2-998-40/PEG/Ion 2 Screen #40 (H04), 185 ml - (0.03 M Citric acid, 0.07 M BIS-TRIS propane)/pH 7.6, 20% w/v Polyethylene glycol 3,350
SDP
Supplier:  Hampton Research HR299840
Encompass
HR2-998-40/PEG/Ion 2 Screen #40 (H04), 185 ml - (0.03 M Citric acid, 0.07 M BIS-TRIS propane)/pH 7.6, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-249-2