Learn More
Hampton Research PEG/ION SCREEN NO05 185ML
SDP
Supplier:  Hampton Research HR292205
Encompass
HR2-922-05/PEG/Ion Screen #05 (A5), 185 ml - 0.2 M Magnesium chloride hexahydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-182-8