Learn More
Hampton Research PEG/ION SCREEN NO06 185ML
SDP
Supplier:  Hampton Research HR292206
Encompass
HR2-922-06/PEG/Ion Screen #06 (A6), 185 ml - 0.2 M Sodium chloride, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-182-9